Learn More
Medchemexpress LLC 1H-Indole-3-methanol, 1-(phenylmethyl)- | 60941-76-6 | 237.30 | 5 MG
SDP
Catalog No. 5000415258
Encompass_Preferred
1-Benzyl-I3C is an E1/E2/E3 enzyme inhibitor, specifically a NEDD4-1 inhibitor, demonstrating significant anticancer activity. It directly inhibits the ubiquitination activity of NEDD4-1 with an IC50 of 12.3μM, which is considerably more potent than its precursor compound I3C. In vitro ubiquitination experiments and thermal stability analysis indicate that 1-Benzyl-I3C binds to the catalytic HECT domain of NEDD4-1. This product is for research use only.
  • Inhibits E1/E2/E3 enzymes
  • Shows significant anticancer activity
  • Binds to the catalytic HECT domain of NEDD4-1
  • High purity of 99.68%
  • White to off-white solid appearance
Catalog No. 50-004-15258 Supplier Medchemexpress LLC Supplier No. HYN101595MG
1-Benzyl-I3C is an E1/E2/E3 enzyme inhibitor, specifically a NEDD4-1 inhibitor, demonstrating significant anticancer activity. It directly inhibits the ubiquitination activity of NEDD4-1 with an IC50 of 12.3μM, which is considerably more potent than its precursor compound I3C. In vitro ubiquitination experiments and thermal stability analysis indicate that 1-Benzyl-I3C binds to the catalytic HECT domain of NEDD4-1. This product is for research use only.
  • Inhibits E1/E2/E3 enzymes
  • Shows significant anticancer activity
  • Binds to the catalytic HECT domain of NEDD4-1
  • High purity of 99.68%
  • White to off-white solid appearance