Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Indole-3-propanoic acid | 830-96-6 | MFCD00005660 | 99.9% | 189.21 g/mol | C11H11NO2 | 1 ML
SDP

Catalog No. 5000303378
Encompass_Preferred

3-Indolepropionic acid is an indole-derived endogenous metabolite and a potent antioxidant investigated for neuroprotective effects and metabolic biomarker research. It scavenges hydroxyl radicals effectively without generating pro-oxidant intermediates and is supplied for research use as analytical standards and solution formulations.

  • Potent antioxidant activity and suggested neuroprotective effects.
  • More potent scavenger of hydroxyl radicals than melatonin.
  • Scavenges radicals without producing reactive pro-oxidant intermediates.
  • Gut microbiota-derived metabolite and potential biomarker for type 2 diabetes.
  • Molecular formula C11H11NO2; molecular weight 189.21 g/mol; CAS 830-96-6.
  • Available in researcher-ready solution formulations (for example, 10 mM in 1 mL) for in vitro use.

Catalog No. 50-003-03378 Supplier Medchemexpress LLC Supplier No. HYW01522910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Indolepropionic acid is an indole-derived endogenous metabolite and a potent antioxidant investigated for neuroprotective effects and metabolic biomarker research. It scavenges hydroxyl radicals effectively without generating pro-oxidant intermediates and is supplied for research use as analytical standards and solution formulations.

  • Potent antioxidant activity and suggested neuroprotective effects.
  • More potent scavenger of hydroxyl radicals than melatonin.
  • Scavenges radicals without producing reactive pro-oxidant intermediates.
  • Gut microbiota-derived metabolite and potential biomarker for type 2 diabetes.
  • Molecular formula C11H11NO2; molecular weight 189.21 g/mol; CAS 830-96-6.
  • Available in researcher-ready solution formulations (for example, 10 mM in 1 mL) for in vitro use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.