Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-4-methanamine | 3468-18-6 | MFCD04973297 | >=98.0% | 146.19 g·mol⁻¹ | C9H10N2 | 250 MG
SDP

Catalog No. 5000208464
Encompass_Preferred

4-Aminomethylindole (NSC 131886) is an organic synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied for use as a building block in drug synthesis and small-molecule preparation.

  • High purity (≥98.0%).
  • CAS number 3468-18-6.
  • Molecular formula C9H10N2.
  • Molecular weight 146.19 g·mol⁻¹.
  • Available in small-scale pack sizes such as 250 MG for research use.

Catalog No. 50-002-08464 Supplier Medchemexpress LLC Supplier No. 250MGHYW046161
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminomethylindole (NSC 131886) is an organic synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied for use as a building block in drug synthesis and small-molecule preparation.

  • High purity (≥98.0%).
  • CAS number 3468-18-6.
  • Molecular formula C9H10N2.
  • Molecular weight 146.19 g·mol⁻¹.
  • Available in small-scale pack sizes such as 250 MG for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.