Learn More
Medchemexpress LLC 1H-indole-5-carboxamide | 1670-87-7 | MFCD07779486 | 99.2% | 160.18 | C9H8N2O | 100 MG
SDP
Catalog No. 5000302578
Encompass_Preferred
SD-169 (1H-indole-5-carboxamide) is a selective, ATP-competitive inhibitor of p38 MAP kinases used as a research reagent for biochemical and cellular studies of MAPK signaling. It exhibits low-nanomolar activity against p38α and is supplied with high reported purity to support reproducible experimental results.
  • Selective inhibitor of p38α and p38β with low-nanomolar potency.
  • Suitable for biochemical and cellular assays.
  • High purity for reproducible results.
  • Available as a solid and as DMSO solution formats.
  • Stable under standard storage conditions for solid and solution.
Catalog No. 50-003-02578 Supplier Medchemexpress LLC Supplier No. HYW015445100MG
SD-169 (1H-indole-5-carboxamide) is a selective, ATP-competitive inhibitor of p38 MAP kinases used as a research reagent for biochemical and cellular studies of MAPK signaling. It exhibits low-nanomolar activity against p38α and is supplied with high reported purity to support reproducible experimental results.
  • Selective inhibitor of p38α and p38β with low-nanomolar potency.
  • Suitable for biochemical and cellular assays.
  • High purity for reproducible results.
  • Available as a solid and as DMSO solution formats.
  • Stable under standard storage conditions for solid and solution.