Learn More
Medchemexpress LLC 1H-Isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-5-methyl- | 40313-92-6 | 99.9% | 272.26 | 25 MG
SDP
Catalog No. 5000349085
Encompass_Preferred
Thalidomide-5-methyl is a cereblon (CRBN) ligand derived from Thalidomide, used for recruiting CRBN protein. It can be utilized in the synthesis of PROTACs.
  • Thalidomide-based cereblon (CRBN) ligand.
  • Used in the recruitment of CRBN protein.
  • Can be used to synthesize PROTACs.
Catalog No. 50-003-49085 Supplier Medchemexpress LLC Supplier No. 25MGHY139548
Thalidomide-5-methyl is a cereblon (CRBN) ligand derived from Thalidomide, used for recruiting CRBN protein. It can be utilized in the synthesis of PROTACs.
  • Thalidomide-based cereblon (CRBN) ligand.
  • Used in the recruitment of CRBN protein.
  • Can be used to synthesize PROTACs.