Learn More
Medchemexpress LLC 1H-Isoindole-1,3(2H)-dione, 4-[[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-, hydrochloride (1:1) | 2446474-09-3 | 96.6% | 50 MG
SDP
Catalog No. 5000342575
Encompass_Preferred
Pomalidomide-PEG3-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide based cereblon ligand and a 3-unit PEG linker, commonly used in PROTAC technology. This product is intended for research use only and has not been fully validated for medical applications.
  • E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Utilizes a 3-unit PEG linker
  • Used in PROTAC technology
  • Targets Cereblon
Catalog No. 50-003-42575 Supplier Medchemexpress LLC Supplier No. HY128716B50MG
Pomalidomide-PEG3-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide based cereblon ligand and a 3-unit PEG linker, commonly used in PROTAC technology. This product is intended for research use only and has not been fully validated for medical applications.
  • E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Utilizes a 3-unit PEG linker
  • Used in PROTAC technology
  • Targets Cereblon