Learn More
Medchemexpress LLC 1H-Isoindole-1,3(2H)-dione, 4-[(6-azidohexyl)oxy]-2-(2,6-dioxo-3-piperidinyl)- | 2411389-65-4 | 99.10% | 399.40 | 25 MG
SDP
Catalog No. 5000363444
Encompass_Preferred
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. It functions as a click chemistry reagent, containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules that have Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits intracellular ubiquitin-proteasome system for selective target protein degradation
Catalog No. 50-003-63444 Supplier Medchemexpress LLC Supplier No. 25MGHY148556
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. It functions as a click chemistry reagent, containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules that have Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits intracellular ubiquitin-proteasome system for selective target protein degradation