Learn More
Medchemexpress LLC 1H-Isoindole-1,3(2H)-dione, 5-[2-(2-chloroethoxy)ethoxy]-2-(2,6-dioxo-3-piperidinyl)- | 2230956-57-5 | 96.6% | 380.78 | 25 MG
SDP
Catalog No. 5000350669
Encompass_Preferred
Thalidomide-5-PEG2-Cl is a thalidomide-based cereblon ligand that facilitates the recruitment of CRBN protein. It can be linked to a target protein ligand to create PROTACs, which are designed to harness the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Functions as a thalidomide-based cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a protein ligand via a linker to form PROTACs
  • PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-50669 Supplier Medchemexpress LLC Supplier No. 25MGHY139543
Thalidomide-5-PEG2-Cl is a thalidomide-based cereblon ligand that facilitates the recruitment of CRBN protein. It can be linked to a target protein ligand to create PROTACs, which are designed to harness the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Functions as a thalidomide-based cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a protein ligand via a linker to form PROTACs
  • PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins