Learn More
Medchemexpress LLC 1H-isoindole-4-carboxylic acid, 2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo | 1547163-38-1 | MFCD32063448 | 98.0% | 302.24 g·mol-1 | C14H10N2O6 | 500 MG
SDP
Catalog No. 5000280814
Encompass_Preferred
Thalidomide acid is a thalidomide-derived cereblon (CRBN) ligand bearing a carboxylic acid functional group, supplied as a solid research reagent. It is intended for use as an E3 ligase recruiting moiety in PROTAC design and related chemical biology and synthetic applications.
  • Ligand for E3 ligase (CRBN) used in targeted protein degradation research.
  • Carboxylic acid handle enables amide coupling and linker attachment.
  • High purity (98.0%) suitable for analytical and synthetic work.
  • Solid powder form for convenient handling and formulation.
  • Molecular weight 302.24 g·mol-1; formula C14H10N2O6.
  • Identified by CAS number 1547163-38-1 for unambiguous chemical lookup.
Catalog No. 50-002-80814 Supplier Medchemexpress LLC Supplier No. HYW241635500MG
Thalidomide acid is a thalidomide-derived cereblon (CRBN) ligand bearing a carboxylic acid functional group, supplied as a solid research reagent. It is intended for use as an E3 ligase recruiting moiety in PROTAC design and related chemical biology and synthetic applications.
  • Ligand for E3 ligase (CRBN) used in targeted protein degradation research.
  • Carboxylic acid handle enables amide coupling and linker attachment.
  • High purity (98.0%) suitable for analytical and synthetic work.
  • Solid powder form for convenient handling and formulation.
  • Molecular weight 302.24 g·mol-1; formula C14H10N2O6.
  • Identified by CAS number 1547163-38-1 for unambiguous chemical lookup.