Learn More
AdipoGen 1H-Naphthotriazol
SDP
Supplier:  AdipoGen CDXN0051M005
Encompass_Preferred
Chemical. CAS 269-12-5. Formula C10H7N3. MW 169.2. Synthetic. Fluorescent probe. Used for the detection of nitrite, nitric oxide release and S-nitrosothiols RSNOs. Activatable DNA cleaving agent.
Catalog No. 50-256-0431
 Restricted Item