Learn More
Medchemexpress LLC 1H-oxadiazolo[4,3-a]quinoxalin-1-one | 41443-28-1 | MFCD00792620 | 99.8% | 187.16 g/mol | C9H5N3O2 | 100 MG
SDP
Catalog No. 5000212723
Encompass_Preferred
ODQ (1H-oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase used to probe nitric oxide-sGC signaling and related cellular responses. Supplied as a high-purity research reagent for biochemical and cellular studies.
  • Potent and selective inhibitor of soluble guanylyl cyclase (sGC).
  • Useful for studying nitric oxide-sGC signaling pathways.
  • Reported to enhance cisplatin-induced apoptosis in mesothelioma cells.
  • Provided as a high-purity reagent suitable for biochemical assays.
  • Available in multiple laboratory pack sizes for experimental flexibility.
Catalog No. 50-002-12723 Supplier Medchemexpress LLC Supplier No. HY101255100MG
ODQ (1H-oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase used to probe nitric oxide-sGC signaling and related cellular responses. Supplied as a high-purity research reagent for biochemical and cellular studies.
  • Potent and selective inhibitor of soluble guanylyl cyclase (sGC).
  • Useful for studying nitric oxide-sGC signaling pathways.
  • Reported to enhance cisplatin-induced apoptosis in mesothelioma cells.
  • Provided as a high-purity reagent suitable for biochemical assays.
  • Available in multiple laboratory pack sizes for experimental flexibility.