Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-pyrazole-1-carboximidamide hydrochloride | 4023-02-3 | MFCD00210087 | 98.8% | 146.58 g/mol | C4H7ClN4 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW017032100G

Encompass_Preferred

1H-pyrazole-1-carboximidamide hydrochloride is an organic biochemical reagent used as a building block and assay reagent in life-science research. Supplied as the hydrochloride salt, it is used in organic synthesis, analytical standards, and assay development.

  • High purity suitable for research applications.
  • Molecular weight 146.58 g/mol.
  • CAS number 4023-02-3.
  • Available in 25 g and 100 g pack sizes.
  • Useful as a synthetic intermediate and analytical reference standard.

Catalog No. 50-003-01940


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.