Learn More
Medchemexpress LLC 1H-pyrazole-1-carboximidamide hydrochloride | 4023-02-3 | MFCD00210087 | 98.8% | 146.58 g/mol | C4H7ClN4 | 25 G
SDP
Catalog No. 5000302679
Encompass_Preferred
1H-pyrazole-1-carboximidamide hydrochloride is a small-molecule biochemical reagent provided as the hydrochloride salt for research applications. It functions as a building block and reagent in organic synthesis and peptide guanylation workflows and is supplied with batch-specific purity and documentation.
  • Hydrochloride salt supplied as a solid.
  • Molecular formula C4H7ClN4; molecular weight 146.58 g/mol.
  • Used as a building block or reagent in organic synthesis and guanylation of amines.
  • Available in laboratory-scale quantities (e.g., 25 g, 100 g).
  • Batch-reported purity approximately 98.8% with higher selectable batches.
  • COA, SDS, and datasheet provided for quality and handling information.
Catalog No. 50-003-02679 Supplier Medchemexpress LLC Supplier No. 25GHYW017032
1H-pyrazole-1-carboximidamide hydrochloride is a small-molecule biochemical reagent provided as the hydrochloride salt for research applications. It functions as a building block and reagent in organic synthesis and peptide guanylation workflows and is supplied with batch-specific purity and documentation.
  • Hydrochloride salt supplied as a solid.
  • Molecular formula C4H7ClN4; molecular weight 146.58 g/mol.
  • Used as a building block or reagent in organic synthesis and guanylation of amines.
  • Available in laboratory-scale quantities (e.g., 25 g, 100 g).
  • Batch-reported purity approximately 98.8% with higher selectable batches.
  • COA, SDS, and datasheet provided for quality and handling information.