Learn More
Medchemexpress LLC 1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester | 1026682-51-8 | 237.30 | C12H19N3O2 | 25 MG
SDP
Catalog No. 5000240568
Encompass_Preferred
1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a small-molecule drug intermediate used in medicinal chemistry research. It functions as a PROTAC CDK9-binding moiety and serves as a building block in the synthesis of CDK9-targeting and other active compounds. Supplied as a solid in small research sizes, it is intended for laboratory use in compound development and synthesis.
  • Used as a building block for PROTAC and CDK9-targeted compound synthesis.
  • Supplied as a solid suitable for standard laboratory handling.
  • Molecular formula C12H19N3O2 and molecular weight 237.30 g·mol⁻¹.
  • Available in small research sizes, including 25 MG.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-40568 Supplier Medchemexpress LLC Supplier No. HY10744825MG
Add to Cart
Add to Cart
1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a small-molecule drug intermediate used in medicinal chemistry research. It functions as a PROTAC CDK9-binding moiety and serves as a building block in the synthesis of CDK9-targeting and other active compounds. Supplied as a solid in small research sizes, it is intended for laboratory use in compound development and synthesis.
  • Used as a building block for PROTAC and CDK9-targeted compound synthesis.
  • Supplied as a solid suitable for standard laboratory handling.
  • Molecular formula C12H19N3O2 and molecular weight 237.30 g·mol⁻¹.
  • Available in small research sizes, including 25 MG.
  • For research use only; not for human or diagnostic use.