Learn More
Medchemexpress LLC 1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester | 1026682-51-8 | 237.30 g/mol | C12H19N3O2 | 100 MG
SDP
Catalog No. 5000239747
Encompass_Preferred
1H-Pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a drug intermediate used in the synthesis of CDK9-targeting PROTACs and other active small molecules. Chemical identifiers: CAS 1026682-51-8; formula C12H19N3O2; molecular weight 237.30 g/mol.
  • Drug intermediate for synthesis of PROTACs and other small molecules.
  • Pyrazole-containing ester functional group enables conjugation chemistry.
  • Defined molecular formula and weight support analytical characterization.
  • Intended for research and medicinal chemistry applications.
Catalog No. 50-002-39747 Supplier Medchemexpress LLC Supplier No. HY107448100MG
1H-Pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a drug intermediate used in the synthesis of CDK9-targeting PROTACs and other active small molecules. Chemical identifiers: CAS 1026682-51-8; formula C12H19N3O2; molecular weight 237.30 g/mol.
  • Drug intermediate for synthesis of PROTACs and other small molecules.
  • Pyrazole-containing ester functional group enables conjugation chemistry.
  • Defined molecular formula and weight support analytical characterization.
  • Intended for research and medicinal chemistry applications.