Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-pyrazole, 1-(methyl-d3)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) | 1022151-50-3 | ≥98.0% | 211.08 | C10H14D3BN2O2 | 5 G
SDP

Catalog No. 5000280575
Encompass_Preferred

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled synthetic intermediate supplied for research use. It is provided as a colorless to light yellow solid and is characterized by high purity and defined isotopic enrichment for use in labeling and analytical applications.

  • Purity ≥98.0%.
  • Deuterium-labeled for isotope tracing and mass-shift experiments.
  • Suitable as a synthetic intermediate in organic synthesis.
  • Colorless to light yellow solid with defined physical properties.
  • Molecular formula C10H14D3BN2O2; molecular weight 211.08.

Catalog No. 50-002-80575 Supplier Medchemexpress LLC Supplier No. HYW2495145G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled synthetic intermediate supplied for research use. It is provided as a colorless to light yellow solid and is characterized by high purity and defined isotopic enrichment for use in labeling and analytical applications.

  • Purity ≥98.0%.
  • Deuterium-labeled for isotope tracing and mass-shift experiments.
  • Suitable as a synthetic intermediate in organic synthesis.
  • Colorless to light yellow solid with defined physical properties.
  • Molecular formula C10H14D3BN2O2; molecular weight 211.08.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.