Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-pyrazole, 1-(methyl-d3)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- | 1022151-50-3 | ≥98.0% | 211.08 g/mol | C10H14D3BN2O2 | 100 MG
SDP

Catalog No. 5000281295
Encompass_Preferred

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled boronic ester supplied as a high-purity research reagent for use as an isotope-labeled building block, tracer, and analytical standard in synthetic and analytical chemistry. It is intended for method development, quantitative analysis, and incorporation into cross-coupling workflows.

  • Deuterium-labeled (d3) boronic ester suitable as tracer and internal standard.
  • Purity ≥98.0%.
  • Molecular formula C10H14D3BN2O2; molecular weight 211.08 g/mol.
  • Available in small-scale packages such as 100 mg.
  • Supplied with COA and SDS for batch-specific documentation.
  • Functional pinacol boronate enables Suzuki-Miyaura cross-coupling.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS method development.

Catalog No. 50-002-81295 Supplier Medchemexpress LLC Supplier No. HYW249514100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled boronic ester supplied as a high-purity research reagent for use as an isotope-labeled building block, tracer, and analytical standard in synthetic and analytical chemistry. It is intended for method development, quantitative analysis, and incorporation into cross-coupling workflows.

  • Deuterium-labeled (d3) boronic ester suitable as tracer and internal standard.
  • Purity ≥98.0%.
  • Molecular formula C10H14D3BN2O2; molecular weight 211.08 g/mol.
  • Available in small-scale packages such as 100 mg.
  • Supplied with COA and SDS for batch-specific documentation.
  • Functional pinacol boronate enables Suzuki-Miyaura cross-coupling.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.