Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Pyrazole-1-(N-methylcarboxamidine) hydrochloride, 96%, Thermo Scientific™

Catalog No. AAH3060714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3060714 25 g
AAH3060706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3060714 Supplier Thermo Scientific Chemicals Supplier No. H3060714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194852-88-5
Molecular Formula C5H9ClN4
Molecular Weight (g/mol) 160.605
MDL Number MFCD11114408
InChI Key SYSWHVMZAHBYJO-UHFFFAOYSA-N
Synonym 1h-pyrazole-1-n-methylcarboxamidine hydrochloride, n'-methylpyrazole-1-carboximidamide hydrochloride, e-n'-methylpyrazole-1-carboximidamide hydrochloride, n1-methyl-1h-pyrazole-1-carboxamidine hydrochloride
PubChem CID 73995114
IUPAC Name N'-methylpyrazole-1-carboximidamide;hydrochloride
SMILES CN=C(N)N1C=CC=N1.Cl

Specifications

Melting Point 156°C to 159°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 160.61
Percent Purity 96%
Chemical Name or Material 1H-Pyrazole-1-(N-methylcarboxamidine) hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.