Learn More
Medchemexpress LLC 1H-pyrazole | 288-13-1 | 99.8% | C3H4N2 | 10 G
SDP
Catalog No. 5000434675
Encompass_Preferred
1H-pyrazole is a five-membered heterocyclic compound. Its derivatives are orally effective antimalarial and antileishmanial agents that can potentially modulate targets like alcohol dehydrogenase and NMDA receptors, making this compound useful in research related to malaria and leishmaniasis.
  • Five-membered heterocyclic compound
  • Derivatives act as orally effective antimalarial and antileishmanial agents
  • Potential to modulate alcohol dehydrogenase and NMDA receptors
  • Inhibits Plasmodium berghei in infected mice
  • Inhibits promastigotes of Leishmania aethiopica
Catalog No. 50-004-34675 Supplier Medchemexpress LLC Supplier No. HY7622810G
1H-pyrazole is a five-membered heterocyclic compound. Its derivatives are orally effective antimalarial and antileishmanial agents that can potentially modulate targets like alcohol dehydrogenase and NMDA receptors, making this compound useful in research related to malaria and leishmaniasis.
  • Five-membered heterocyclic compound
  • Derivatives act as orally effective antimalarial and antileishmanial agents
  • Potential to modulate alcohol dehydrogenase and NMDA receptors
  • Inhibits Plasmodium berghei in infected mice
  • Inhibits promastigotes of Leishmania aethiopica