Learn More
Medchemexpress LLC 1H-pyrazole | 288-13-1 | 99.8% | C3H4N2 | 25 G
SDP
Catalog No. 5000357618
Encompass_Preferred
1H-pyrazole is a five-membered heterocyclic compound. Its derivatives are orally effective antimalarial and antileishmanial agents with the potential to modulate targets such as alcohol dehydrogenase and NMDA receptors. It can be used in research related to malaria and leishmaniasis.
  • Inhibitory effects on *Plasmodium berghei* in infected mice
  • Inhibitory effects on promastigotes of *Leishmania aethiopica*
Catalog No. 50-003-57618 Supplier Medchemexpress LLC Supplier No. 25GHY76228
1H-pyrazole is a five-membered heterocyclic compound. Its derivatives are orally effective antimalarial and antileishmanial agents with the potential to modulate targets such as alcohol dehydrogenase and NMDA receptors. It can be used in research related to malaria and leishmaniasis.
  • Inhibitory effects on *Plasmodium berghei* in infected mice
  • Inhibitory effects on promastigotes of *Leishmania aethiopica*