Learn More
Medchemexpress LLC 1H-pyrazole, 3,4-dimethyl-, phosphate (1:1) | 202842-98-6 | MFCD09842544 | 98.0% | 194.13 g/mol | C5H11N2O4P | 100 G
SDP
Catalog No. 5000301470
Encompass_Preferred
3,4-Dimethylpyrazole phosphate (3,4-DMPP) is the phosphate salt of 3,4-dimethyl-1H-pyrazole used as a nitrification inhibitor and research chemical. Supplied as a solid with a typical purity of 98%, it is used in soil science and fertilizer formulation to reduce the microbial conversion of ammonium to nitrate and limit nitrogen losses.
  • Acts as a nitrification inhibitor, reducing microbial conversion of ammonium to nitrate.
  • High purity (~98%) suitable for analytical and formulation work.
  • Supplied as a stable solid for ease of handling and storage.
  • Available in multiple pack sizes to support laboratory and scale-up use.
  • Batch certificate of analysis available for quality traceability.
Catalog No. 50-003-01470 Supplier Medchemexpress LLC Supplier No. HYW019334100G
3,4-Dimethylpyrazole phosphate (3,4-DMPP) is the phosphate salt of 3,4-dimethyl-1H-pyrazole used as a nitrification inhibitor and research chemical. Supplied as a solid with a typical purity of 98%, it is used in soil science and fertilizer formulation to reduce the microbial conversion of ammonium to nitrate and limit nitrogen losses.
  • Acts as a nitrification inhibitor, reducing microbial conversion of ammonium to nitrate.
  • High purity (~98%) suitable for analytical and formulation work.
  • Supplied as a stable solid for ease of handling and storage.
  • Available in multiple pack sizes to support laboratory and scale-up use.
  • Batch certificate of analysis available for quality traceability.