Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrazole, 3-methyl- | 1453-58-3 | 99.5% | C4H6N2 | 10 G
SDP

Catalog No. 5000434954
Encompass_Preferred

3-Methylpyrazole is a chemical compound primarily used as a nitrification inhibitor in soil. It has also shown the ability to inhibit the metabolism of MAM by rat liver microsomes in in vitro studies. This compound is supplied with high purity, making it suitable for various research and laboratory applications.

  • Used as a nitrification inhibitor in soil
  • Inhibits the metabolism of MAM by rat liver microsomes
  • High purity of 99.5%
  • Molecular weight of 82.11
  • Appearance varies from solid to liquid based on temperature
  • Light yellow to reddish brown in color
  • Soluble in water and PBS

Catalog No. 50-004-34954 Supplier Medchemexpress LLC Supplier No. HY6605410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methylpyrazole is a chemical compound primarily used as a nitrification inhibitor in soil. It has also shown the ability to inhibit the metabolism of MAM by rat liver microsomes in in vitro studies. This compound is supplied with high purity, making it suitable for various research and laboratory applications.

  • Used as a nitrification inhibitor in soil
  • Inhibits the metabolism of MAM by rat liver microsomes
  • High purity of 99.5%
  • Molecular weight of 82.11
  • Appearance varies from solid to liquid based on temperature
  • Light yellow to reddish brown in color
  • Soluble in water and PBS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.