Learn More
Medchemexpress LLC 1H-Pyrazole, 3-methyl- | 1453-58-3 | MFCD00005240 | 99.5% | C4H6N2 | 10 MM 1 ML
SDP
Catalog No. 5000433573
Encompass_Preferred
3-Methylpyrazole is used as a nitrification inhibitor in soil and can inhibit the metabolism of MAM by rat liver microsomes. This product is provided in a solution format.
  • Used as a nitrification inhibitor
  • Inhibits metabolism of MAM by rat liver microsomes
  • High purity of 99.5%
  • Light yellow to reddish brown appearance
Catalog No. 50-004-33573 Supplier Medchemexpress LLC Supplier No. HY6605410MM/1ML
3-Methylpyrazole is used as a nitrification inhibitor in soil and can inhibit the metabolism of MAM by rat liver microsomes. This product is provided in a solution format.
  • Used as a nitrification inhibitor
  • Inhibits metabolism of MAM by rat liver microsomes
  • High purity of 99.5%
  • Light yellow to reddish brown appearance