Learn More
Medchemexpress LLC 1H-Pyrazole, 3-nitro- | 26621-44-3 | 113.08 | 25 G
SDP
Catalog No. 5000449461
Encompass_Preferred
3-Nitropyrazole is a biochemical reagent and nitrogen-containing heterocyclic compound used as a biological material or organic compound for life science research. It can also be utilized in the synthesis of other active compounds.
  • Biochemical reagent for life science research
  • Organic compound for life science research
  • Nitrogen-containing heterocyclic compound
  • Used in the synthesis of other active compounds
  • Nitro group readily undergoes nucleophilic substitution reactions
Catalog No. 50-004-49461 Supplier Medchemexpress LLC Supplier No. 25GHYW002457
3-Nitropyrazole is a biochemical reagent and nitrogen-containing heterocyclic compound used as a biological material or organic compound for life science research. It can also be utilized in the synthesis of other active compounds.
  • Biochemical reagent for life science research
  • Organic compound for life science research
  • Nitrogen-containing heterocyclic compound
  • Used in the synthesis of other active compounds
  • Nitro group readily undergoes nucleophilic substitution reactions