Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrazole-4-carboxaldehyde, 3-iodo-1-(tetrahydro-2H-pyran-2-yl)- | 1627924-19-9 | 99.8% | 306.10 | 250 MG
SDP

Catalog No. 5000402282
Encompass_Preferred

1H-Pyrazole-4-carboxaldehyde, 3-iodo-1-(tetrahydro-2H-pyran-2-yl)- is an off-white to light yellow solid. It has a molecular weight of 306.10 and a purity of 99.83%. This chemical is designed for research use.

  • Off-white to light yellow solid appearance
  • Purity of 99.83%
  • Store at -20°C, protect from light, stored under nitrogen
  • For research use only

Catalog No. 50-004-02282 Supplier Medchemexpress LLC Supplier No. 250MGHY41514
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1H-Pyrazole-4-carboxaldehyde, 3-iodo-1-(tetrahydro-2H-pyran-2-yl)- is an off-white to light yellow solid. It has a molecular weight of 306.10 and a purity of 99.83%. This chemical is designed for research use.

  • Off-white to light yellow solid appearance
  • Purity of 99.83%
  • Store at -20°C, protect from light, stored under nitrogen
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.