Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H-Pyrazole-5-boronic acid

Catalog No. CDS014043 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014043 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014043 Supplier Sigma Aldrich Supplier No. CDS014043250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl perfluorooctanoate (mepfoa) is reported to be formed during storage of perfluorooctanoic acid (pfoa) in methanol. its physical properties like density, boiling point and refractive index have been determined experimentally.methyl perfluorooctanoate may be employed in the following studies:
  • Synthesis of monodisperse perfluoroalkyl n-polyethoxylated amides, potential nonionic fluorinated surfactants
  • To investigate the mutual solubility curves of fluorocarbon-hydrocarbon systems
  • Synthesis of fluorine-containing monomer, n-(5-hydroxypent-1-yl)perfluorooctaneamide
  • To investigate the vinyl ether formulations for use in step and flash imprint lithography
  • Preparation of 2-chloro-1,1-dicyano-2-(perfluoroheptyl)ethylene, a precursor for synthesizing perfluorocarbon-soluble dyes

Specifications

Molecular Formula C3H5BN2O2
Linear Formula C3H5BN2O2
Quantity 5 mL
MDL Number MFCD07368247
Molecular Weight (g/mol) 111.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.