Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 1H-Pyrazole-5-carbaldehyde, 3920-50-1, MFCD06657574, 1g

Supplier:  Matrix Scientific 0322731G

Encompass_Preferred

Molecular Formula C4H4N2O, Purity 95%, Molecular Weight 96.09, Melting Point 142-144°, Boiling Point 300

Catalog No. 50-277-118


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.