Learn More
Medchemexpress LLC 1H-pyrazolo[3,4-d]pyrimidin-4-amine, N,1-diphenyl- | 68380-53-0 | 99.5% | 287.32 Da | C17H13N5 | 10 MG
SDP
Catalog No. 5000289259
Encompass_Preferred
Antifungal agent 112 (Compound 1e) is a purified small-molecule research compound that shows potent in vitro antifungal activity against Candida albicans, Aspergillus niger, and Aspergillus fumigatus (reported MICs 0.0024, 0.0022, and 0.0028 M, respectively). It is supplied as a high-purity powder, soluble in DMSO, and intended for laboratory research use.
  • High purity suitable for research use.
  • Potent in vitro activity against Candida and Aspergillus species.
  • Soluble in DMSO at the concentrations described by the supplier.
  • Stable as a powder when stored under recommended conditions.
  • Available in small milligram pack sizes for laboratory experiments.
Catalog No. 50-002-89259 Supplier Medchemexpress LLC Supplier No. HY16462010MG
Antifungal agent 112 (Compound 1e) is a purified small-molecule research compound that shows potent in vitro antifungal activity against Candida albicans, Aspergillus niger, and Aspergillus fumigatus (reported MICs 0.0024, 0.0022, and 0.0028 M, respectively). It is supplied as a high-purity powder, soluble in DMSO, and intended for laboratory research use.
  • High purity suitable for research use.
  • Potent in vitro activity against Candida and Aspergillus species.
  • Soluble in DMSO at the concentrations described by the supplier.
  • Stable as a powder when stored under recommended conditions.
  • Available in small milligram pack sizes for laboratory experiments.