Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-pyrrole-1-acetic acid, 2,5-dihydro-2,5-dioxo- | 25021-08-3 | MFCD00457254 | 100.0% | 155.11 g/mol | C6H5NO4 | 50 MG
SDP

Catalog No. 5000302039
Encompass_Preferred

2-Maleimido acetic acid is a maleimide-functionalized carboxylic acid used as a reactive intermediate in organic synthesis and bioconjugation. It reacts selectively with thiol groups, enabling targeted conjugation workflows, and is supplied as a solid for laboratory use.

  • Reactive maleimide group enables selective thiol conjugation.
  • High purity (99.97%) suitable for analytical and synthetic use.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 155.11 g/mol and formula C6H5NO4.
  • Storage stability under recommended cold conditions for extended shelf life.

Catalog No. 50-003-02039 Supplier Medchemexpress LLC Supplier No. HYW01783150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Maleimido acetic acid is a maleimide-functionalized carboxylic acid used as a reactive intermediate in organic synthesis and bioconjugation. It reacts selectively with thiol groups, enabling targeted conjugation workflows, and is supplied as a solid for laboratory use.

  • Reactive maleimide group enables selective thiol conjugation.
  • High purity (99.97%) suitable for analytical and synthetic use.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 155.11 g/mol and formula C6H5NO4.
  • Storage stability under recommended cold conditions for extended shelf life.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.