Learn More
Medchemexpress LLC 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, calcium salt, hydrate (2:1:3), (βR,δR)- | 344423-98-9 | 99.8% | 605.71 | 1 ML
SDP
Catalog No. 5000436307
Encompass_Preferred
Atorvastatin calcium salt trihydrate is a statin-class drug used to lower blood cholesterol levels. It is effective in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides by inhibiting HMG-CoA reductase, an enzyme responsible for cholesterol production in the liver. This product also serves as a biochemical reagent for life science research.
  • Controls high levels of LDL cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Inhibits HMG-CoA reductase.
  • Useful as a biochemical reagent for life science research.
Catalog No. 50-004-36307 Supplier Medchemexpress LLC Supplier No. HY11249010MM/1ML
Atorvastatin calcium salt trihydrate is a statin-class drug used to lower blood cholesterol levels. It is effective in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides by inhibiting HMG-CoA reductase, an enzyme responsible for cholesterol production in the liver. This product also serves as a biochemical reagent for life science research.
  • Controls high levels of LDL cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Inhibits HMG-CoA reductase.
  • Useful as a biochemical reagent for life science research.