Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrrole-2,5-dione, 1-[2-(2-hydroxyethoxy)ethyl]- | 34321-81-8 | 99.7% | C8H11NO4 | 250 MG
SDP

Catalog No. 5000352599
Encompass_Preferred

Mal-PEG2-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase to a ligand for the target protein via a linker.

  • Molecular weight of 185.18
  • Purity of 99.7%
  • Appearance as a colorless to off-white solid-liquid mixture
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure

Catalog No. 50-003-52599 Supplier Medchemexpress LLC Supplier No. 250MGHY140968
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG2-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase to a ligand for the target protein via a linker.

  • Molecular weight of 185.18
  • Purity of 99.7%
  • Appearance as a colorless to off-white solid-liquid mixture
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.