Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1h-pyrrole-2,5-dione, 1-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)- | 1262681-30-0 | 99.7% | 311.33 | 25 MG
SDP

Catalog No. 5000348731
Encompass_Preferred

Mal-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
  • Appearance: Colorless to light yellow oil

Catalog No. 50-003-48731 Supplier Medchemexpress LLC Supplier No. 25MGHY140037
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
  • Appearance: Colorless to light yellow oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.