Learn More
Medchemexpress LLC 1H-Pyrrole-2,5-dione, 1-(4-methylphenyl)- | 1631-28-3 | ≥98.0% | 10 MG
SDP
Catalog No. 5000339976
Encompass_Preferred
p-Tolylmaleimide is a naphthalimide derivative with cytotoxic effects on cancer cells. It has shown the ability to arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis, making it a subject of interest in oncology research.
  • Cytotoxic effects on cancer cells
  • Arrests cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase
  • Induces apoptosis
  • Inhibits human acute myeloid leukemia cell K562
  • Inhibits human acute myeloid leukemia cell Jurkat
  • Appears as a light yellow to yellow solid
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-39976 Supplier Medchemexpress LLC Supplier No. HY12959210MG
p-Tolylmaleimide is a naphthalimide derivative with cytotoxic effects on cancer cells. It has shown the ability to arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis, making it a subject of interest in oncology research.
  • Cytotoxic effects on cancer cells
  • Arrests cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase
  • Induces apoptosis
  • Inhibits human acute myeloid leukemia cell K562
  • Inhibits human acute myeloid leukemia cell Jurkat
  • Appears as a light yellow to yellow solid
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light