Learn More
Medchemexpress LLC 1H-Pyrrole-2-carboxaldehyde | 1003-29-8 | 100.0% | C5H5NO | 1 ML
SDP
Catalog No. 5000434631
Encompass_Preferred
Pyrrole-2-carboxaldehyde, also known as 2-Formylpyrrole, is a five-membered N-heterocyclic system. It exhibits a dimeric form in the solid phase and a monomeric form in solution. This compound features a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It typically appears as a white to light brown solid.
  • Available as a 10 mM solution in DMSO.
  • Exhibits dimeric form in solid phase.
  • Exhibits monomeric form in solution.
  • Contains a pyrrole group as a proton donor.
  • Contains a carbonyl group as a proton acceptor.
  • Store solid at 4°C, protected from light and under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • For research use only.
Catalog No. 50-004-34631 Supplier Medchemexpress LLC Supplier No. HY7781710MM/1ML
Pyrrole-2-carboxaldehyde, also known as 2-Formylpyrrole, is a five-membered N-heterocyclic system. It exhibits a dimeric form in the solid phase and a monomeric form in solution. This compound features a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It typically appears as a white to light brown solid.
  • Available as a 10 mM solution in DMSO.
  • Exhibits dimeric form in solid phase.
  • Exhibits monomeric form in solution.
  • Contains a pyrrole group as a proton donor.
  • Contains a carbonyl group as a proton acceptor.
  • Store solid at 4°C, protected from light and under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • For research use only.