Learn More
Medchemexpress LLC 1H-pyrrole-2-carboxamide, N-cyclopropyl-3-methyl-4-[[6-(trifluoromethyl)-3-pyridinyl]methyl]- | 2426616-55-7 | 99.7% | C16H16F3N3O | 10MG
SDP
Catalog No. 5000259814
Encompass_Preferred
DSM502 is a pyrrole-based dihydroorotate dehydrogenase (DHODH) inhibitor selective for Plasmodium DHODH. It shows nanomolar potency in enzyme and cell assays, and has demonstrated in vivo antimalarial efficacy in mouse models. Supplied as a high-purity solid or DMSO solution, it is intended for research use.
  • Pyrrole-based DHODH inhibitor selective for Plasmodium enzymes.
  • Nanomolar potency in enzyme and cell-based assays.
  • Demonstrated efficacy in mouse infection models.
  • High purity suitable for biochemical studies.
  • Soluble at high concentration in DMSO for in vitro use.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-59814 Supplier Medchemexpress LLC Supplier No. HY13217010MG
DSM502 is a pyrrole-based dihydroorotate dehydrogenase (DHODH) inhibitor selective for Plasmodium DHODH. It shows nanomolar potency in enzyme and cell assays, and has demonstrated in vivo antimalarial efficacy in mouse models. Supplied as a high-purity solid or DMSO solution, it is intended for research use.
  • Pyrrole-based DHODH inhibitor selective for Plasmodium enzymes.
  • Nanomolar potency in enzyme and cell-based assays.
  • Demonstrated efficacy in mouse infection models.
  • High purity suitable for biochemical studies.
  • Soluble at high concentration in DMSO for in vitro use.
  • Stable when stored as powder at -20°C for long-term storage.