Learn More
Medchemexpress LLC 1H-Pyrrole-2-carboxylic acid | 634-97-9 | 111.10 | 100 G
SDP
Catalog No. 5000446430
Encompass_Preferred
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae sp. Nov*. It is a biological form of pyrrole that is commonly found in natural compounds and serves as a precursor for various pyrrole derivatives in biosynthetic processes.
  • Natural alkaloid
  • Biological form of pyrrole
  • Precursor for pyrrole derivatives
  • Exhibits antineuroinflammatory activity
Catalog No. 50-004-46430 Supplier Medchemexpress LLC Supplier No. HYW001963100G
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae sp. Nov*. It is a biological form of pyrrole that is commonly found in natural compounds and serves as a precursor for various pyrrole derivatives in biosynthetic processes.
  • Natural alkaloid
  • Biological form of pyrrole
  • Precursor for pyrrole derivatives
  • Exhibits antineuroinflammatory activity