Learn More
Medchemexpress LLC 1H-Pyrrole-2-carboxylic acid | 634-97-9 | MFCD00005219 | 111.10 | 10 G
SDP
Catalog No. 5000445955
Encompass_Preferred
Pyrrole-2-carboxylic acid is a natural alkaloid derived from the marine bacterium *Pelomonas puraquae sp. Nov*. It represents a biological form of pyrrole commonly found in natural compounds. This substance serves as a precursor in biosynthetic processes for various pyrrole derivatives, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin.
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Store in solvent at -80°C for up to 6 months or at -20°C for up to 1 month.
  • For solutions in water, dilute to working solution, then filter and sterilize with a 0.22 μm filter prior to use.
  • For *in vivo* experiments, prepare working solutions freshly and use them on the same day.
  • Exercise caution in protocol selection if the continuous dosing period extends beyond half a month.
Catalog No. 50-004-45955 Supplier Medchemexpress LLC Supplier No. HYW00196310G
Pyrrole-2-carboxylic acid is a natural alkaloid derived from the marine bacterium *Pelomonas puraquae sp. Nov*. It represents a biological form of pyrrole commonly found in natural compounds. This substance serves as a precursor in biosynthetic processes for various pyrrole derivatives, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin.
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Store in solvent at -80°C for up to 6 months or at -20°C for up to 1 month.
  • For solutions in water, dilute to working solution, then filter and sterilize with a 0.22 μm filter prior to use.
  • For *in vivo* experiments, prepare working solutions freshly and use them on the same day.
  • Exercise caution in protocol selection if the continuous dosing period extends beyond half a month.