Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrrole-3-carboxaldehyde | 7126-39-8 | 99.9% | C5H5NO | 5 G
SDP

Catalog No. 5000434404
Encompass_Preferred

1H-Pyrrole-3-carboxaldehyde is a biochemical reagent suitable for life science-related research. It functions as a biological material or organic compound, intended for research use only and not for medical applications.

  • High purity of 99.9%
  • Appears as an off-white to light brown solid
  • Molecular formula is C5H5NO
  • Molecular weight is 95.10
  • Store at 4°C under nitrogen

Catalog No. 50-004-34404 Supplier Medchemexpress LLC Supplier No. HY593075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1H-Pyrrole-3-carboxaldehyde is a biochemical reagent suitable for life science-related research. It functions as a biological material or organic compound, intended for research use only and not for medical applications.

  • High purity of 99.9%
  • Appears as an off-white to light brown solid
  • Molecular formula is C5H5NO
  • Molecular weight is 95.10
  • Store at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.