Learn More
Medchemexpress LLC 1H-pyrrole-3-carboxamide, 1-(methylsulfonyl)-N-[(1S)-3-(methylthio)-1-[[[4-[3-(4-pyridinyl)phenyl] | 2368903-18-6 | 98.4% | C25H25N5O4S3 | 10MG
SDP
Catalog No. 5000258303
Encompass_Preferred
FHT-1015 is a potent, selective small-molecule inhibitor of SMARCA4 and SMARCA2 ATPase activity, with low-nanomolar IC50 values. Supplied as a white to off-white solid and as a 10 mM solution in DMSO, the compound is intended for in vitro epigenetics and chromatin-remodeling research and includes manufacturer guidance for storage to maintain stability.
  • High potency against SMARCA4 and SMARCA2, IC50 ≈ 4-5 nM.
  • High chemical purity (>98%).
  • Available as solid and DMSO solution formats for experimental flexibility.
  • White to off-white solid appearance, suitable for analytical handling.
  • Storage recommendations provided for long- and short-term stability.
Catalog No. 50-002-58303 Supplier Medchemexpress LLC Supplier No. HY14489610MG
FHT-1015 is a potent, selective small-molecule inhibitor of SMARCA4 and SMARCA2 ATPase activity, with low-nanomolar IC50 values. Supplied as a white to off-white solid and as a 10 mM solution in DMSO, the compound is intended for in vitro epigenetics and chromatin-remodeling research and includes manufacturer guidance for storage to maintain stability.
  • High potency against SMARCA4 and SMARCA2, IC50 ≈ 4-5 nM.
  • High chemical purity (>98%).
  • Available as solid and DMSO solution formats for experimental flexibility.
  • White to off-white solid appearance, suitable for analytical handling.
  • Storage recommendations provided for long- and short-term stability.