Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrrole-3-carboxamide, N-[6-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoin | 2222635-92-7 | 99.3% | 654.69 | 100 MG
SDP

Catalog No. 5000352358
Encompass_Preferred

MI-389 is a PROTAC translation termination factor GSPT1 degrader. It disrupts a target that is a shared dependency in different AML and ALL cell lines, and its action is dependent on the CRL4CRBN E3 ligase.

  • Disrupts GSPT1 target
  • Action dependent on CRL4CRBN E3 ligase
  • Target is Cereblon
  • EC50 value of 21.3 nM on Kasumi 1 cells
  • Antiproliferative activity against human Kasumi 1 cells
  • Molecular formula: C35H35FN6O6
  • Appearance: Yellow to orange solid
  • Purity: 99.34%
  • Solubility: 66.67 mg/mL (101.83 mM) in DMSO
  • Storage: 4°C, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Catalog No. 50-003-52358 Supplier Medchemexpress LLC Supplier No. HY136688100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MI-389 is a PROTAC translation termination factor GSPT1 degrader. It disrupts a target that is a shared dependency in different AML and ALL cell lines, and its action is dependent on the CRL4CRBN E3 ligase.

  • Disrupts GSPT1 target
  • Action dependent on CRL4CRBN E3 ligase
  • Target is Cereblon
  • EC50 value of 21.3 nM on Kasumi 1 cells
  • Antiproliferative activity against human Kasumi 1 cells
  • Molecular formula: C35H35FN6O6
  • Appearance: Yellow to orange solid
  • Purity: 99.34%
  • Solubility: 66.67 mg/mL (101.83 mM) in DMSO
  • Storage: 4°C, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.