Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrrolo[2,3-b]pyridine 7-oxide | 55052-24-9 | 99.2% | C7H6N2O | 10 G
SDP

Catalog No. 5000358805
Encompass_Preferred

1H-Pyrrolo[2,3-b]pyridine 7-oxide is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active pharmaceutical compounds. It typically appears as an off-white to pale purple solid, with a high purity of 99.2%.

  • Used as a drug intermediate
  • Suitable for synthesis of various active compounds
  • Appears as a solid
  • Color: Off-white to pale purple
  • Molecular weight: 134.14
  • For research use only

Catalog No. 50-003-58805 Supplier Medchemexpress LLC Supplier No. HY5051710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1H-Pyrrolo[2,3-b]pyridine 7-oxide is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active pharmaceutical compounds. It typically appears as an off-white to pale purple solid, with a high purity of 99.2%.

  • Used as a drug intermediate
  • Suitable for synthesis of various active compounds
  • Appears as a solid
  • Color: Off-white to pale purple
  • Molecular weight: 134.14
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.