Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Tetrazole 98%, Thermo Scientific™

Catalog No. AC168280250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168280250 25 g
AC168280050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168280250 Supplier Thermo Scientific Chemicals Supplier No. 168280250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-94-8
Molecular Formula CH2N4
Molecular Weight (g/mol) 70.055
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
PubChem CID 67519
ChEBI CHEBI:33193
IUPAC Name 2H-tetrazole
SMILES C1=NNN=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 155°C to 158°C
Color White
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 26, 346
Synonym 1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
Formula Weight 70.04
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Tetrazole, 98%
Show More Show Less
EINECSNumber 206-023-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.