Learn More
Medchemexpress LLC 1H-Thieno[3,4-d]imidazole-4-pentanamide, hexahydro-2-oxo-N-[6-[[1-oxo-3-(2-pyridinyldithio)propyl] | 129179-83-5 | 99.7% | 539.78 | 50 MG
SDP
Catalog No. 5000347959
Encompass_Preferred
Biotin-HPDP is a biochemical reagent that can couple with GMPS and label free protein thiols. It is suitable for use as a biological material or organic compound for life science related research.
  • Couples with GMPS
  • Labels free protein thiols
  • Modified surfaces adsorb proteins
Catalog No. 50-003-47959 Supplier Medchemexpress LLC Supplier No. HY13676950MG
Biotin-HPDP is a biochemical reagent that can couple with GMPS and label free protein thiols. It is suitable for use as a biological material or organic compound for life science related research.
  • Couples with GMPS
  • Labels free protein thiols
  • Modified surfaces adsorb proteins