Learn More
Medchemexpress LLC 1H-Thieno[3,4-d]imidazole-4-pentanamide, N-[19-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)- | 1255942-07-4 | 99.49% | 50 MG
SDP
Catalog No. 5000340280
Encompass_Preferred
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that contains a biotin group and four PEG units. It functions as a versatile biotinylation reagent, enabling the introduction of a biotin moiety to azide-labeled biomolecules. This process occurs through a copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction. It is a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Azadibenzocyclooctyne-biotin derivative
  • Contains a biotin group and 4 PEGs
  • Versatile biotinylation reagent
  • Used for introducing a biotin moiety to azide-labeled biomolecules
  • Utilizes copper-free strain-promoted alkyne-azide click chemistry (SPAAC)
  • Contains a DBCO group for SPAAC with azide groups
  • For research use only
Catalog No. 50-003-40280 Supplier Medchemexpress LLC Supplier No. HY13080950MG
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that contains a biotin group and four PEG units. It functions as a versatile biotinylation reagent, enabling the introduction of a biotin moiety to azide-labeled biomolecules. This process occurs through a copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction. It is a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Azadibenzocyclooctyne-biotin derivative
  • Contains a biotin group and 4 PEGs
  • Versatile biotinylation reagent
  • Used for introducing a biotin moiety to azide-labeled biomolecules
  • Utilizes copper-free strain-promoted alkyne-azide click chemistry (SPAAC)
  • Contains a DBCO group for SPAAC with azide groups
  • For research use only