Learn More
Medchemexpress LLC 1H-Thieno[3,4-d]imidazole-4-pentanamide, N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethyl]hexahydro-2-oxo- | 875770-34-6 | ≥97.0% | 25 MG
SDP
Catalog No. 5000342761
Encompass_Preferred
Biotin-PEG3-azide is a PEG-based PROTAC linker and a click chemistry reagent. It contains an azide group, allowing it to participate in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions. This compound can be conjugated to antiviral inhibitors and demonstrates broad-spectrum inhibition against various pathogenic viruses.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Can be conjugated to antiviral inhibitors
  • Demonstrates broad-spectrum inhibition against various pathogenic viruses
Catalog No. 50-003-42761 Supplier Medchemexpress LLC Supplier No. 25MGHY130143
Biotin-PEG3-azide is a PEG-based PROTAC linker and a click chemistry reagent. It contains an azide group, allowing it to participate in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions. This compound can be conjugated to antiviral inhibitors and demonstrates broad-spectrum inhibition against various pathogenic viruses.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Can be conjugated to antiviral inhibitors
  • Demonstrates broad-spectrum inhibition against various pathogenic viruses