Learn More
Medchemexpress LLC 1H-Thieno[3,4-d]imidazole-4-pentanamide, N-[2-[2-(2-azidoethoxy)ethoxy]ethyl] hexahydro-2-oxo- | 1910803-72-3 | 97.0% | 100 MG
SDP
Catalog No. 5000342988
Encompass_Preferred
Biotin-PEG2-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-42988 Supplier Medchemexpress LLC Supplier No. HY126957100MG
Biotin-PEG2-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions