Learn More
Medchemexpress LLC 1H-thieno[3,4-d]imidazole-4-pentanamide, N-[4-[(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)methyl] | 188682-72-6 | MFCD00213997 | 99.9% | 533.68 g·mol⁻¹ | C26H39N5O5S | 10 MG
SDP
Catalog No. 5000288114
Encompass_Preferred
Biotin-BMCC is a maleimide-activated biotinylation reagent that reacts selectively with free thiol groups of cysteines to install a biotin tag for detection, affinity capture, or purification of proteins and peptides.
  • Reacts selectively with free thiol groups (cysteines).
  • Introduces a biotin handle compatible with streptavidin/avidin affinity methods.
  • High chemical purity (99.9%).
  • Suitable for labeling proteins and peptides for detection and affinity purification.
Catalog No. 50-002-88114 Supplier Medchemexpress LLC Supplier No. HY15905710MG
Biotin-BMCC is a maleimide-activated biotinylation reagent that reacts selectively with free thiol groups of cysteines to install a biotin tag for detection, affinity capture, or purification of proteins and peptides.
  • Reacts selectively with free thiol groups (cysteines).
  • Introduces a biotin handle compatible with streptavidin/avidin affinity methods.
  • High chemical purity (99.9%).
  • Suitable for labeling proteins and peptides for detection and affinity purification.