Learn More
Cayman Chemical 1Heptadecnoylracglycerol 10mg
SDP
Catalog No. 502938607
Encompass_Preferred
A monoacylglycerol; active against various bacteria (MIC = 78 UG/ml for all); has been found in T. africana, I. sonorae, and wheat bran
Catalog No. 50-293-8607 Supplier Cayman Chemical Supplier No. 2818010 MG
A monoacylglycerol; active against various bacteria (MIC = 78 UG/ml for all); has been found in T. africana, I. sonorae, and wheat bran