Learn More
Cayman Chemical (1H-Indol-3-yl)(2,2,3,3-tetramethylcyclopropyl)methanone, Purity: ≥98%, CAS: 895152-66-6, 10 mg
SDP
Catalog No. 502895431
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-289-5431 Supplier Cayman Chemical Supplier No. 900196610 MG
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications