Learn More
Chemglass Life Sciences 1" Labeling Tape Rainbow Pack, 2 Rolls of Each Color: White, Yellow, Green, Red, Orange, Blue and Lavender
SDP
Catalog No. 501531651
Encompass_Preferred
1" Labeling Tape Rainbow Pack, 2 Rolls of Each Color: White, Yellow, Green, Red, Orange, Blue and Lavender. Labeling tape for glass or plastic. Each rainbow pack includes two rolls of each color (white, yellow, green, red, orange, blue and lavender), with the same size cores. Adheres to any dry surface. Removes cleanly and easily. Durable, waterproof, oil and acid-resistant. Withstands temperatures from -23 to 121C. Accepts permanent marker, pencil or pen. 500" rolls, 1 inch wide
Catalog No. 50-153-1651 Supplier Chemglass Life Sciences Supplier No. CG4035R100
1" Labeling Tape Rainbow Pack, 2 Rolls of Each Color: White, Yellow, Green, Red, Orange, Blue and Lavender. Labeling tape for glass or plastic. Each rainbow pack includes two rolls of each color (white, yellow, green, red, orange, blue and lavender), with the same size cores. Adheres to any dry surface. Removes cleanly and easily. Durable, waterproof, oil and acid-resistant. Withstands temperatures from -23 to 121C. Accepts permanent marker, pencil or pen. 500" rolls, 1 inch wide